Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ TrkA Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 156.25 pg/mL |
---|---|
Assay Sensitivity | 156.25-10000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009721
|
Novus Biologicals™
NBP27934115PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human TrkA.Specifications
156.25 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
NTRK1 | |
Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase | |
TrkA |
156.25-10000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Human TrkA. | |
Sandwich ELISA | |
4914 | |
Rabbit/Mouse | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10000 pg/mL is recommended. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title