Learn More
Alfa Aesar™ TrkA Inhibitor
Manufacturer:  Alfa Aesar™ J64810LB0
 View more versions of this product
Catalog No. AAJ64810LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
2°C to 8°C
TrkA Inhibitor
388626-12-8
315.40
Light Sensitive
Yellow
Solid
An oxindole compound that acts as a cell-permeable, reversible, potent, and highly selective inhibitor of TrkA
C15H13N3O3S
(Z)-4-(((2-Oxoindolin-3-ylidene)methyl)amino)benzenesulfonamide