Learn More
TrkA Mouse anti-Human, Alexa Fluor™ 405, Clone: 994307, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB1752V100UG
Catalog No. FB1752V100U
Add to cart
This item is not returnable. View return policy
Description
Description
TrkA Monoclonal antibody specifically detects TrkA in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TrkA
994307
Flow Cytometry 0.25-1 μg/106 cells
P04629
Mouse
IgG1
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human TrkA in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 405
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, NTRK1, NTRK-1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A
Mouse myeloma cell line NS0-derived recombinant human TrkA, Ala33-Glu407, Accession No. P04629
100 μg
Primary
4914
Human