Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TrkA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586240
Description
Purity is > 95% by SDS-PAGE.
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
Specifications
TrkA | |
Polyclonal | |
Unconjugated | |
Ntrk1 | |
C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; Trka; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P04629 | |
Ntrk1 | |
Synthetic peptide, corresponding to amino acids 751-796 of Human Trk A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction