Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkA Rabbit anti-Human, Clone: RM423, Novus Biologicals™

Rabbit Monoclonal Antibody

$104.00 - $366.50

Specifications

Antigen TrkA
Clone RM423
Dilution Western Blot 1:1000-1:4000, Immunohistochemistry 1:100-1:400
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB119700
View Documents
Novus Biologicals
NBP30917625UL
25 μg
Each for $104.00
Add to cart
 
NB119699
View Documents
Novus Biologicals
NBP309176100UL
100 μg
Each for $366.50
Add to cart
 
Description

Description

TrkA Monoclonal antibody specifically detects TrkA in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

TrkA
Western Blot 1:1000-1:4000, Immunohistochemistry 1:100-1:400
Unconjugated
Rabbit
Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase
PBS, 50% Glycerol, 1% BSA
4914
IgG
Primary
Store at -20°C.
RM423
Western Blot, Immunohistochemistry
Purified
RUO
Human
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A
A peptide corresponding to the C-terminus of human TrkA
Monoclonal
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit