Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkA Rabbit anti-Human, DyLight 594, Clone: 24, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289782DL594

Catalog No. NB109232


Add to cart

Description

Description

TrkA Monoclonal specifically detects TrkA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TrkA
24
50 mM Sodium Borate
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A
This antibody was obtained from a rabbit immunized with purified, recombinant Human TrkA (NP_002520.2; Met 1-Pro 382).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 594
Purified
Rabbit
IgG
0.1 mL
Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase
Primary
4914
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit