Learn More
Novus Biologicals™ TrkB Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2794345PLATES
 View more versions of this product
Catalog No. NB009908
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TrkB.
Specifications
Specifications
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TrkB.
10.98 pg/mL
4915
RUO
TrkB
GPCR, Neuroendocrinology, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction
Human
NTRK2
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.3 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 700 pg/mL is recommended.
Sandwich ELISA
Antibody Pairs
Sandwich ELISA
  • Standard: Each vial contains 28 ng of recombinant Human TrkB.
  • Capture Antibody: 1 mg/mL of mouse anti-Human TrkB monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human TrkB monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Mouse
    5 Plates
    10.98-700 pg/mL
    BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B