Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkC Polyclonal Antibody, Invitrogen™

Sheep Polyclonal Antibody

Supplier:  Thermo Scientific OST00119W

Catalog No. PIOST00119W

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μl of sterile water.

a tyrosine-protein kinase receptor for neurotrophin-3.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TrkC
Polyclonal
NTRK3
Neurotrophic tyrosine kinase receptor type 3, TrkC tyrosine kinase, GP145-TrkC, Trk-C, NT-3 growth factor receptor, NTRK3
Sheep
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Q16288, Q6VNS1, Q03351
NTRK3
A synthetic peptide from the extracellular domain of rat TrkC.
RUO
18213, 29613, 4916
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit