Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkC Rat anti-Mouse, DyLight 680, Clone: 6G17, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11094150FR

Catalog No. NBT94150FR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TrkC
6G17
Western Blot
Purified
NTRK3
Recombinant Mouse TrkC extracellular domain
Protein G purified
Cancer, Phospho Specific, Protein Kinase
Primary
4916
Mouse
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This recognizes mouse TrkC in western blot. It has no cross-reactivity with mouse TrkB.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit