Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRMT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559438
Description
Immunogen sequence: EHCGQRHQLG GPMWAEPIHD LDFVGRVLEA VSANPGRFHT SERIRGVLSV ITEELPDVPL YYTLDQLSST IHCNTP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
TRMT1 is a tRNA(m(2)(2)G(26))dimethyltransferase.
Specifications
TRMT1 | |
Polyclonal | |
Unconjugated | |
TRMT1 | |
6720406L13Rik; 6720477L24; D8Ertd812e; FLJ20244; im:7150454; LOC553298 protein; n(2),N(2)-dimethylguanosine tRNA methyltransferase; N(2),N(2)-dimethylguanosine tRNA methyltransferase; tRNA (guanine(26)-N(2))-dimethyltransferase; RGD1305992; TRM1; TRM1 tRNA methyltransferase 1; TRM1 tRNA methyltransferase 1 homolog; TRMT1; tRNA (guanine(26)-N(2))-dimethyltransferase; tRNA 2,2-dimethylguanosine-26 methyltransferase; tRNA methyltransferase 1; tRNA methyltransferase 1 homolog; tRNA methyltransferase 1 homolog (S. cerevisiae); tRNA(guanine-26,N(2)-N(2)) methyltransferase; tRNA(m(2,2)G26)dimethyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55621 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NXH9 | |
TRMT1 | |
Recombinant protein corresponding to Human TRMT1. Recombinant protein control fragment (Product #RP-98375). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction