Learn More
TRMT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149701AP
Catalog No. 50-172-9875
Add to cart
Description
Description
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.
Specifications
Specifications
TRMT1
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3TX08, Q9NXH9
TRM1, TRMT1
Rabbit
IgG
150 μL
-20°C
Primary
212528, 288914, 55621
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TRMT1
Liquid
Trmt1
TRMT1 Fusion Protein Ag6837
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat