Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRMT2B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548138

Catalog No. PA548138

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probable S-adenosyl-L-methionine-dependent methyltransferase that catalyzes the formation of 5-methyl-uridine at position 54 (m5U54) in all tRNA. May also have a role in tRNA stabilization or maturation.
Specifications

Specifications

TRMT2B
Polyclonal
PBS with 0.09% sodium azide
Q96GJ1
tRNA (uracil(54)-C(5))-methyltransferase homolog, TRM2 homolog, TRMT2B, CXorf34
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79979
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TRMT2B
Liquid
TRMT2B
KLH conjugated synthetic peptide between 136-165 amino acids from the N-terminal region of human TRMT2B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit