Learn More
Aviva Systems Biology Corp TRMT5 Antibody - middle region (ARP48868_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48868_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRMT5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRMT5. Alternate names: KIAA1393, MGC111453, TRM5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-59944