Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRMT61B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543226
Description
Peptide sequence: PLSISDIGTG CLSSLENLRL PTLREESSPR ELEDSSGDQG RCGPTHQGSE Sequence homology: Human: 100%.
Methyltransferase that catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in various tRNAs in mitochondrion, including tRNA(Leu) (decephering codons UUA or UUG), tRNA(Lys) and tRNA(Ser) (decephering codons UCA, UCU, UCG or UCC).
Specifications
TRMT61B | |
Polyclonal | |
Unconjugated | |
TRMT61B | |
1110039F03Rik; 623451; Gm6431; mRNA methyladenosine-N(1)-methyltransferase; potential tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61B; potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B; TRMT61B; tRNA (adenine(58)-N(1))-methyltransferase, mitochondrial; tRNA methyltransferase 61 homolog B; tRNA methyltransferase 61B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55006 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BVS5 | |
TRMT61B | |
Synthetic peptide directed towards the N-terminal of human FLJ20628 (aa 71-120). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction