Learn More
Aviva Systems Biology Corp TRMU Antibody - C-terminal region (ARP48819_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48819_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRMU. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRMU. Alternate names: MGC99627, MTO2, MTU1, TRMT, TRMT1, TRNT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59928