Troemner™ Alloy 8 Metric Precision Individual Weights, UltraClass with Statement of Accuracy

Excellent for high-precision calibrations and reference standards

Manufacturer: TroemnerSupplier Diversity Partner 70130

 View more versions of this product

Catalog No. 02-215-99

  Check Availability
Add to Cart

    Specifications

    Includes Polycarbonate case
    Quantity Each
    Material Alloy 8 Stainless Steel
    Density 8.03g/cm3 at 20°C
    Product Type Two piece metric analytical weight
    Certifications/Compliance Statement of accuracy, ISO9001
    Classification UltraClass
    Packaging Quantity 1
    Weight (Metric) 1kg
    Mass 1kg
    Shape Cylindrical with handling knob

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit