Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Alloy 8 Metric Precision Individual Weights, UltraClass

Excellent for high-precision calibrations and reference standards

Manufacturer:  Troemner™ 70280

 View more versions of this product

Catalog No. 02-214M


Add to cart

Includes: Forceps

Description

Description

Low magnetic susceptibility, uniform consistency and high resistance to corrosion. Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.

  • Low magnetic susceptibility, uniform consistency and high resistance to corrosion.
  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
  • Weights 1mg through 2kg are supplied in a high quality polycarbonate case

Certifications

A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements

Specifications

Specifications

One-piece metric analytical sheet weight
Stainless Steel
7.95g/cm3 at 20°C
Flat with one end turned up for easy handling
Each
UltraClass
200mg
200mg
Polycarbonate case
1
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit