Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 1 with NVLAP Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer:  Troemner™ 72171W
 View more versions of this product
Catalog No. 02-100-183
Add to cart
Description
Description
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
  • Low magnetic susceptibility, uniform consistency and high resistance to corrosion
  • Used in extremely stable environments
  • 1 to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • Weights sets 1mg-500g are provided in a high-quality polcarbonate set case
  • Weight sets where the largest weight is 1kg and above are supplied in a felt line oak case
  • Each polycarbonate case is placed in its own individual compartment
Order Info
Sets are supplied in polypropylene case with individual compartments for each weight.
Specifications
Specifications
Large metric analytical weight set
ASTM Class 1
1mg to 1kg
Stainless Steel cylindrical weight: 1kg, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case in Wood case
NVLAP, ISO9001
Stainless Steel, Aluminum
20kg
25/Set