Troemner™ Alloy 8 Metric Precision Weight Sets, Class 1 with Statement of Accuracy

Excellent for high-precision calibrations and reference standards

Manufacturer: TroemnerSupplier Diversity Partner 72311

 View more versions of this product

Catalog No. 02-319H

  Check Availability
Add to Cart

    Specifications

    Includes Cylindrical weight: 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, Polycarbonate case
    Classification ASTM Class 1
    Quantity 16/Set
    Packaging Quantity Multiple
    Weight (Metric) 10mg to 50g
    Product Type Medium metric analytical weight set
    Certifications/Compliance Statement of accuracy, ISO9001
    Material Stainless Steel
    Mass 50g, (2)20g, 10g, 5g, (2)2g, 1g, 500mg, (2)200mg, 100mg, 50mg, (2)20mg, 10mg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit