Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 1
Excellent for high-precision calibrations and reference standards
Manufacturer:  Troemner™ 72141
 View more versions of this product
Catalog No. 02-216-011
Add to cart
Includes: Forceps
Description
Description
Manufactured from a specially developed austenitic stainless-stee—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless-steel.
  • Each fractional weight is embossed with its value
  • Weights from 1 to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
  • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
  • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
  • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with “2” (2mg, 20mg, etc.) and one weight for all other inclusive values
Certifications
Statement of Accuracy
Specifications
Specifications
Large metric analytical weight set
Stainless Steel
2kg, 1kg, 500g, (2)200g, 100g, 50g, (2)20g, 10g, 5g, (2)2g, 1g
14/Set
ASTM Class 1
1g to 2kg
Cylindrical weight: 2kg, 1kg, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case in Wood case