Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 3
Excellent for high-precision calibrations and reference standards
Manufacturer:  Troemner™ 72283
 View more versions of this product
Catalog No. 02-215-20C
Includes: Forceps
Description
Description
Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless-steel.
  • Each fractional weight is embossed with its value
  • Weights from 1 to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
  • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
  • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
  • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with “2” (2mg, 20mg, etc.) and one weight for all other inclusive values
Specifications
Specifications
Medium metric analytical weight set
Stainless Steel, Aluminum
1mg to 100g
21/Set
ASTM Class 3
1mg to 100g
Stainless Steel cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Multiple