Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 4 with Statement of Accuracy
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer:  Troemner™ 72244
 View more versions of this product
Catalog No. 02-215-51B
  Check Availability
Add to cart
Description
Description
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
  • Used in extremely stable environments
  • 1 to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • Weight sets 1mg-500g are provided in a high quality polycarbonate set case
  • Weight sets where the largest weight is 1kg and above are supplied in a felt line oak case
  • Each polycarbonate case is placed in its own individual compartment
Certifications
A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements
Specifications
Specifications
Medium metric analytical weight set
ASTM Class 4
1g to 100g
Cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case
Multiple
Statement of accuracy, ISO9001
Stainless Steel
100g, 50g, (2)20g, 10g, 5g, (2)2g, 1g
9/Set