Troemner™ Alloy 8 Metric Precision Weight Sets, Class 4 with Traceable Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer: TroemnerSupplier Diversity Partner 7221-4T
 View all options for this product
Catalog No. 02-214-102
  Check Availability
Add to Cart
    Product Type Medium metric analytical weight set
    Certifications/Compliance Traceable certificate, ISO9001
    Material Alloy 8 Stainless Steel
    Weight (Metric) 1mg to 200g
    Mass (2)200g, 100g, 50g, (2)20g, 10g, 5g, (2)2g, 1g, 500mg, (2)200mg, 100mg, 50mg, (2)20mg, 10mg, 5mg, (2)2mg, 1mg
    Includes Stainless Steel cylindrical weight: 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case
    Quantity 23/Set
    Packaging Quantity Multiple
    Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
    • Used in extreme stable environments
    • 1 to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
    • Weight sets 1mg-500g are provided in a high quality polycarbonate set case
    • Weight sets where the largest weight is 1kg and above are supplied in a felt lined oak case with each polycarbonate case placed in its own individual compartment
  • Description & Specifications
Related Products
Product Content Feedback