Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, UltraClass with NVLAP Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer:  Troemner™ 72230W
 View more versions of this product
Catalog No. 02-214-135
  Check Availability
Add to cart
Description
Description
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
  • Used in extremely stable environments
  • 1 to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • Weight sets 1mg-500g are provided in a high-quality polycarbonate set case
  • Weight sets where the largest weight is 1kg and above are supplied in a felt line oak case
  • Each polycarbonate case is placed in its own individual compartment
Specifications
Specifications
Medium metric analytical weight set
UltraClass
1mg to 200g
Stainless Steel cylindrical weight: 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Multiple
NVLAP, ISO9001
Aluminum
200g, 100g, 50g, (2)20g, 10g, 5g, (2)2g, 1g, 500mg, (2)200mg, 100mg, 50mg, (2)20mg, 10mg, 5mg, (2)2mg, 1mg
22/Set