Troemner™ Alloy 8 Metric Precision Weight Sets, UltraClass with NVLAP Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer: TroemnerSupplier Diversity Partner 72170W
 View more versions of this product
Catalog No. 02-100-169
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Large metric analytical weight set
    Includes Stainless Steel cylindrical weight: 1kg, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case in Wood case
    Quantity 25/Set
    Certifications/Compliance NVLAP, ISO9001
    Classification UltraClass
    Material Stainless Steel, Aluminum
    Weight (Metric) 1mg to 1kg
    Mass 1kg, 500g, (2) 200g, 100g, 50g, (2) 20g, 10g, 5g, (2) 2g, 1g, Stainless Steel sheet weight: 500mg, (2) 200mg, 100mg, 50mg, (2) 20mg, 10mg, 5mg, Aluminium sheet weight: (2) 2mg, 1mg
  • Description & Specifications
Related Products
Product Content Feedback