Troemner™ Alloy™ 8 Metric Precision Weight Sets, UltraClass with Statement of Accuracy
Excellent for high-precision calibrations and reference standards
Manufacturer: TroemnerSupplier Diversity Partner 72400
 View more versions of this product
Catalog No. 02-100-161
  Check Availability
Add to Cart
    Specifications
    Product Type Small metric analytical weight set
    Certifications/Compliance Statement of accuracy, ISO9001
    Includes Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case
    Classification UltraClass
    Quantity 12/Set
    Material Stainless Steel, Aluminum
    Weight (Metric) 1mg to 500g
    Mass 1 to 500mg
  • Description & Specifications
Related Products
Product Content Feedback