Troemner™ Alloy™ 8 Metric Precision Weight Sets, UltraClass with Statement of Accuracy

Excellent for high-precision calibrations and reference standards

Manufacturer: TroemnerSupplier Diversity Partner 7231-0

 View all options for this product

Catalog No. 02-100-171

  Check Availability
Add to Cart


    Product Type Medium metric analytical weight set
    Certifications/Compliance Statement of accuracy, ISO9001
    Material Stainless Steel
    Weight (Metric) 10mg to 50g
    Mass 10mg to 50g
    Includes Cylindrical weight: 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, Polycarbonate case
    Quantity 16/Set

    Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless steel.

    • Each fractional weight is embossed with its value
    • Weights from 1mg to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
    • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
    • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
    • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with “2”quotation (2mg, 20mg, etc.) and one weight for all other inclusive values
    • Each weight set (100g to 1mg) is packaged in a polypropylene case
    • Weight sets 500g and above come packaged in a wood case
    • All weights come with a statement of accuracy

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit