Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, UltraClass with Traceable Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer:  TroemnerSupplier Diversity Partner 72170T
 View more versions of this product
Catalog No. 02-100-170
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Large metric analytical weight set
UltraClass
1mg to 1kg
Stainless Steel cylindrical weight: 1kg, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case in Wood case
Traceable certificate, ISO9001
Stainless Steel, Aluminum
1mg to 1kg
25/Set