Learn More
Troemner™ Cal-Paks™ Weight Sets: Class 0
Highly polished Alloy 8 stainless-steel weights
Manufacturer:  Troemner™ 725000W
 View more versions of this product
Catalog No. 02-215-600
  Check Availability
Add to cart
Includes: Master carrying case; three individually cased precision weights; USB Flashdrive with electronic NVLAP™ certification; NVLAP Lab Code 105013-0; NVLAP certificate; and accessories for proper handling and weight care
Description
Description
ANSI™/ASTM™ Class 0.
  • Components and case are linked through the use of a unique serial number
  • Clear plastic case
  • Individual weights in high-quality polycarbonate cases
Compliance
Meet ISO™/IEC 17025 requirements.
Specifications
Specifications
Small Weight Set
UltraClass
1mg, 200mg, 2g
3/Set
NVLAP, ISO9001
2.2g
Troemner Alloy 8 cylindrical weight: 2g, Stainless Steel weight: 200mg, Aluminuml weight: 1mg, Polycarbonate case; NVLAP certificate
3