Troemner™ Cal-Paks™ Weight Sets: Class 1

Highly polished Alloy 8 stainless-steel weights

Manufacturer: TroemnerSupplier Diversity Partner 72611W

 View more versions of this product

Catalog No. 02-215-611

  Check Availability
Add to Cart

    Specifications

    Includes Cylindrical weight: 500g, 50g, 2g, Polycarbonate case in Polypropylene case; NVLAP certificate
    Classification ASTM Class 1
    Product Type Cal-Paks™ large weight set
    Certifications/Compliance NVLAP, ISO9001
    Quantity 3/Set
    Packaging Quantity 3
    Weight (Metric) 552g
    Density 7.95g/cm3 at 20°C
    Mass 2g, 50g, 500g

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit