Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troemner™ Individual Analytical Precision Weights, Class 1 with Traceable Certificate
Designed for your most demanding weighing applications
Supplier: Troemner™ 30391575
Description
Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.
- Precision Analytical Individual Weights are made from the finest materials; manufactured in sizes ranging from 1mg through 50kg
- Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
- Fractional weights 3mg and below are made of aluminum
- Weights 1mg to 2kg are supplied in a high-quality polycarbonate case
- Used as reference standards as well as calibrating Class I balances as specified in OIML R 76
- Used to calibrate Class 1 weights
- Calibrate certain Class I and Class II balances and scales as specified in OIML R 76
- Can also be used to calibrate Class 2 weights
Specifications
Metric Analytical Sheet Weight with One Side Turned Up | |
ASTM Class 1 | |
1 mg | |
1 mg | |
Polycarbonate case | |
1 |
Traceable certificate, ISO9001 | |
Aluminum | |
2.7g/cm3 at 20°C | |
Flat with one side turned up | |
Each |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction