Learn More
Troemner Individual Analytical Precision Weights, Class 2 with NVLAP Certificate
Designed for your most demanding weighing applications
Manufacturer:  TroemnerSupplier Diversity Partner 70222W
 View more versions of this product
Catalog No. 02-217-2CC
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Metric analytical weight with handling knob
ASTM Class 2
5g
5g
Polycarbonate case
1
NVLAP, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with handling knob
Each