Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Individual Analytical Precision Weights, Class 3 with Traceable Certificate

Low magnetic susceptibility, uniform consistency and high resistance to corrosion

Manufacturer:  Troemner™ 70203T

 View more versions of this product

Catalog No. 02-215-25ET


Add to cart

Description

Description

Designed for your most demanding weighing applications, Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.

  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
  • Weights 1mg through 2kg are supplied in a high quality polycarbonate case

  • Reference standards as well as calibrating Class I balances as specified in OIML R 76
  • Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g

Order Info

Protective cases that are supplied with weights can be ordered separately.

Specifications

Specifications

Metric analytical weight
ASTM Class 3
20g
20g
Polycarbonate case
1
Traceable certificate, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with handling knob
Each
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit