Learn More
Troemner™ Individual Analytical Precision Weights, Class 3 with Traceable Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer:  TroemnerSupplier Diversity Partner 70173T
 View more versions of this product
Catalog No. 02-215-25GT
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Metric analytical weight
ASTM Class 3
100g
100g
Polycarbonate case
1
Traceable certificate, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with handling knob
Each