Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate
Designed for your most demanding weighing applications
Manufacturer: TroemnerSupplier Diversity Partner 7034-4W
 View all options for this product
Catalog No. 02-225-528
  Check Availability
Add to Cart
    Product Type Metric analytical sheet weight with one side turned up
    Certifications/Compliance NVLAP, ISO9001
    Material 304 Stainless Steel
    Weight (Metric) 5mg
    Density 7.95g/cm3 at 20°C
    Mass 5mg
    Shape Flat with one side turned up
    Includes Polycarbonate case
    Quantity Each
    Packaging Quantity 1
    Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.
    • Precision Analytical Individual Weights are made from the finest materials; manufactured in sizes ranging from 1mg through 50kg
    • Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
    • Fractional weights 3mg and below are made of aluminum
    • Weights 1mg to 2kg are supplied in a high-quality polycarbonate case
    • Reference standards as well as calibrating Class I balances as specified in OIML R 76
    • Calibration of semi-analytical balances and for student use
  • Description & Specifications
Related Products
Product Content Feedback