Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate

Designed for your most demanding weighing applications

Manufacturer: TroemnerSupplier Diversity Partner 70334W

 View more versions of this product

Catalog No. 02-215-60AC

  Check Availability
Add to Cart

    Specifications

    Classification ASTM Class 4
    Material 304 Stainless Steel
    Product Type Metric analytical sheet weight with one side turned up
    Certifications/Compliance NVLAP, ISO9001
    Includes Polycarbonate case
    Quantity Each
    Packaging Quantity 1
    Weight (Metric) 10mg
    Density 7.95g/cm3 at 20°C
    Mass 10mg
    Shape Flat with one side turned up

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit