Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate

Designed for your most demanding weighing applications

Manufacturer: TroemnerSupplier Diversity Partner 7018-4W

 View all options for this product

Catalog No. 02-225-503

  Check Availability
Add to Cart


    Product Type Metric analytical weight with handling knob
    Certifications/Compliance NVLAP, ISO9001
    Material 316 Stainless Steel
    Weight (Metric) 50g
    Density 7.95g/cm3 at 20°C
    Mass 50g
    Shape Cylindrical with handling knob
    Includes Polycarbonate case
    Quantity Each
    Packaging Quantity 1

    Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.

    • Precision Analytical Individual Weights are made from the finest materials; manufactured in sizes ranging from 1mg through 50kg
    • Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
    • Fractional weights 3mg and below are made of aluminum
    • Weights 1mg to 2kg are supplied in a high-quality polycarbonate case

    • Reference standards as well as calibrating Class I balances as specified in OIML R 76
    • Calibration of semi-analytical balances and for student use

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit