Learn More
Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate
Designed for your most demanding weighing applications
Manufacturer:  TroemnerSupplier Diversity Partner 70254W
 View more versions of this product
Catalog No. 02-215-55AC
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Metric analytical weight with handling knob
ASTM Class 4
1g
1g
Polycarbonate case
1
NVLAP, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with handling knob
Each