Learn More
Troemner™ Individual Analytical Precision Weights, Class 4 with Statement of Accuracy
Designed for your most demanding weighing applications
Manufacturer:  Troemner™ 70214
 View more versions of this product
Catalog No. 02-215-55D
  Check Availability
Add to cart
Description
Description
Low magnetic susceptibility, uniform consistency and high resistance to corrosion. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • For calibration of semi-analytical balances and for student use.
  • Weights 1mg through 2kg are supplied in a high quality polycarbonate case
Certifications
A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements
Specifications
Specifications
Two-piece metric analytical weight
ASTM Class 4
10g
10g
Polycarbonate case
1
Statement of accuracy, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with handling knob
Each