Learn More
Troemner™ Individual Analytical Precision Weights, Class 4 with Statement of Accuracy
Designed for your most demanding weighing applications
Manufacturer:  TroemnerSupplier Diversity Partner 70174
 View more versions of this product
Catalog No. 02-215-55G
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Two-piece metric analytical weight
ASTM Class 4
100g
100g
Polycarbonate case
1
Statement of accuracy, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with handling knob
Each