Learn More
Troemner™ Individual Analytical Precision Weights, Class 4 with Traceable Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer:  TroemnerSupplier Diversity Partner 70264T
 View more versions of this product
Catalog No. 02-215-60FT
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
One-piece metric analytical sheet weight
ASTM Class 4
500mg
500mg
Polycarbonate case
1
Traceable certificate, ISO9001
304 Stainless Steel
7.95g/cm3 at 20°C
Flat with one end turned up for easy handling
Each