Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Individual Analytical Precision Weights, UltraClass with NVLAP Certificate

Highly polished kilogram and gram weights

Manufacturer:  Troemner™ 70330W

 View more versions of this product

Catalog No. 02-214-2S


Add to cart

Description

Description

Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage.

  • Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F)
  • Fractional weights 500mg to 50mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are aluminum
  • The UltraClass weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2
  • This class is used as a reference standard for calibrating other reference standards and weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.01mg
  • Weights are supplied with a NIST/NVLAP Weight Calibration Certificate
  • Weights supplied in a rugged, precision-fit, high quality polycarbonate case

Specifications

Specifications

One-piece metric analytical sheet weight
UltraClass
10mg
10mg
Polycarbonate case
1
NVLAP, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Flat with one end turned up for easy handling
Each
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit