Learn More
Troemner Individual Analytical Precision Weights, UltraClass with NVLAP Certificate
Highly polished kilogram and gram weights
$190.00 - $1992.00
Specifications
Certifications/Compliance | NVLAP, ISO9001 |
---|---|
Classification | UltraClass |
Includes | Polycarbonate case |
Quantity | Each |
Catalog Number | Mfr. No. | Product Type | Mass | Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Mass | Shape | Price | Quantity & Availability | |||
02-214-2A
|
Troemner
70170W |
Two-Piece Metric Analytical Weight | 100 g | Cylindrical with Handling Knob |
Each for $279.00
|
|
02-214-2E
|
Troemner
70210W |
Two-Piece Metric Analytical Weight | 10 g | Cylindrical with Handling Knob |
Each for $219.00
|
|
02-214-MW
|
Troemner
70160W |
Two-Piece Metric Analytical Weight | 200 g | Cylindrical with Handling Knob |
Each for $411.00
|
|
02-214-2F
|
Troemner
70220W |
Two-Piece Metric Analytical Weight | 5 g | Cylindrical with Handling Knob |
Each for $219.00
|
|
02-214-2G
|
Troemner
70230W |
Two-Piece Metric Analytical Weight | 3 g | Cylindrical with Handling Knob |
Each for $219.00
|
|
02-214-2J
|
Troemner
70250W |
Two-Piece Metric Analytical Weight | 1 g | Cylindrical with Handling Knob |
Each for $219.00
|
|
02-214-2K
|
Troemner
70260W |
One Piece Metric Analytical Sheet Weight | 500 mg | Flat with one end turned up for easy handling |
Each for $190.00
|
|
02-214-2M
|
Troemner
70280W |
One Piece Metric Analytical Sheet Weight | 200 mg | Flat with one end turned up for easy handling |
Each for $190.00
|
|
02-214-2N
|
Troemner
70290W |
One Piece Metric Analytical Sheet Weight | 100 mg | Flat with one end turned up for easy handling |
Each for $190.00
|
|
02-214-2Q
|
Troemner
70310W |
One Piece Metric Analytical Sheet Weight | 30 mg | Flat with one end turned up for easy handling |
Each for $190.00
|
|
02-214-2T
|
Troemner
70340W |
One Piece Metric Analytical Sheet Weight | 5 mg | Flat with one end turned up for easy handling |
Each for $190.00
|
|
02-214-2V
|
Troemner
70350W |
One Piece Metric Analytical Sheet Weight | 3 mg | Flat with one end turned up for easy handling |
Each for $190.00
|
|
02-214-2W
|
Troemner
70360W |
One Piece Metric Analytical Sheet Weight | 2 mg | Flat with one end turned up for easy handling |
Each for $190.00
|
|
02-214-116
|
Troemner
70100W |
Two-Piece Metric Analytical Weight | 5 kg | Cylindrical with Handling Knob |
Each for $1,992.00
|
|
02-225-500
|
Troemner
70130W |
Two-Piece Metric Analytical Weight | 1 kg | Cylindrical with Handling Knob |
Each for $952.00
|
|
02-225-534
|
Troemner
70140W |
Two-Piece Metric Analytical Weight | 500 g | Cylindrical with Handling Knob |
Each for $474.00
|
|
02-225-535
|
Troemner
70120W |
Two-Piece Metric Analytical Weight | 2 kg | Cylindrical with Handling Knob |
Each for $1,370.00
|
|
02-214-2S
|
Troemner
70330W |
One Piece Metric Analytical Sheet Weight | 10 mg | Flat with one end turned up for easy handling |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
02-214-2L
|
Troemner
70270W |
One Piece Metric Analytical Sheet Weight | 300 mg | Flat with one end turned up for easy handling |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
02-214-2C
|
Troemner
70190W |
Two-Piece Metric Analytical Weight | 30 g | Cylindrical with Handling Knob |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage.
- Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F)
- Fractional weights 500mg to 50mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are aluminum
- The UltraClass weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2
- This class is used as a reference standard for calibrating other reference standards and weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.01mg
- Weights are supplied with a NIST/NVLAP Weight Calibration Certificate
- Weights supplied in a rugged, precision-fit, high quality polycarbonate case
Specifications
NVLAP, ISO9001 | |
UltraClass | |
Polycarbonate case | |
Each |