Learn More
Troemner™ Individual Analytical Precision Weights, UltraClass with Traceable Certificate
Highly polished kilogram and gram weights
Manufacturer:  TroemnerSupplier Diversity Partner 70320T
 View more versions of this product
Catalog No. 02-214-1R
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Metric analytical sheet weight with one side turned up
UltraClass
20mg
20mg
Polycarbonate case
Traceable certificate, ISO9001
304 Stainless Steel
7.95g/cm3 at 20°C
Flat with one side turned up
Each