Troemner™ Individual Analytical Precision Weights, UltraClass with Traceable Certificate
Highly polished kilogram and gram weights
Manufacturer: TroemnerSupplier Diversity Partner 70160T
 View more versions of this product
Catalog No. 02-214-MT
  Check Availability
Add to Cart
    Specifications
    Product Type Metric analytical weight with handling knob
    Certifications/Compliance Traceable certificate, ISO9001
    Material Alloy 8 Stainless Steel
    Weight (Metric) 200g
    Density 8.03g/cm3 at 20°C
    Mass 200g
    Shape Cylindrical with knob
    Includes Polycarbonate case
    Quantity Each
    Packaging Quantity 1
    Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage.
    • Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F)
    • Fractional weights 500mg to 5mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are aluminum
    • The UltraClass weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2
    • This class is used as a reference standard for calibrating other reference standards and weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.01mg
    • Weights come in a rugged, precision-fit, high-quality polycarbonate case
    • Supplied with a Traceable Certificate
  • Description & Specifications
Related Products
Product Content Feedback