Learn More
Troemner™ Metric Economical Individual Calibration Weights, Class 7 with Statement of Accuracy
Individual weights are available from 2kg to 1mg
Manufacturer:  TroemnerSupplier Diversity Partner 61014S
 View more versions of this product
Catalog No. 02-301-112
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Individual metric economical weight
ASTM Class 7
10g
Cylindrical with handling knob
Each
Statement of accuracy, ISO9001
303 Stainless Steel
7.85g/cm3 at 20°C
21 mg
1