Learn More
Troemner™ Overhead Laboratory Mixer
Talboys Model 101 Overhead Mixer is a general purpose, economical and durable mixer with speeds up to 7500 rpm. Mixer is ideal for low viscosity mixing. The model 101 overhead mixer offers low torque for general purpose mixing and has a continuous duty rating.
Manufacturer:  Troemner™ 30531387
Catalog No. 22-214809
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Top mounted Basic Speed ControlCompact and economical for light duty mixing
  • Offers a single shaft, direct drive motor
  • Mixes up to five gallons of water
  • 5 year industry leading warranty
Overhead mixer includes a collet chuck and 8-inch mounting rod with carriage bolt and wrench for mounting motor to support stands. An additional clamp ( 110B or 112 ) is required to attach the motor's mounting rod to a support stand. Support stands and holders are sold separately. Overhead mixers include a 5 year, industry leading limited warranty on parts and labor.
Specifications
Specifications
0.33A
8 in.
5 in.
12.7cm
120V
11.4cm
752°F
UL, cUL, CE
400°C
33.81 oz., 1000mL
20.3cm
500 to 7500rpm
4.5 in.
72 in. 3-wire cord and plug
5 x 4.5 x 8 in. (12.7 x 11.4 x 20.3cm)