Learn More
Troemner™ Replacement Sleeves for Clamps
Sleeves are easily removed for cleaning or replacement
Manufacturer:  Troemner™ 916250
 View more versions of this product
Catalog No. 02-216-256
  Check Availability
Add to cart
Description
Description
  • Available in vinyl or fiberglass
  • Fiberglass sleeves are recommended for all applications above 100°C (212°F)
Specifications
Specifications
Vinyl
Red
Medium 3 prong clamps
Replacement Sleeves