Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 1 with Statement of Accuracy

Precision-crafted of stainless steel, with a highly polished finish

Manufacturer: TroemnerSupplier Diversity Partner 8110

 View more versions of this product

Catalog No. 02-215-7B

  Check Availability
Add to Cart

    Specifications

    Product Type Electronic balance calibration weight with grip handle
    Certifications/Compliance Statement of accuracy, ISO9001
    Classification ASTM Class 1
    Quantity Each
    Packaging Quantity 1
    Material 303 Stainless Steel
    Weight (Metric) 10kg
    Density 7.85g/cm3 at 20°C
    Mass 10kg
    Shape Cylindrical with recessed grip

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit