Learn More
Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 1 with Statement of Accuracy
Precision-crafted of stainless steel, with a highly polished finish
Manufacturer:  TroemnerSupplier Diversity Partner 8102
 View more versions of this product
Catalog No. 02-215-7D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Electronic balance calibration weight with grip handle
ASTM Class 1
25kg
25kg
Each
Statement of accuracy, ISO9001
Stainless Steel
7.85g/cm3 at 20°C
Cylindrical with recessed grip
1